N-((4-Amino-7-(1H-pyrazol-5-yl)-3H-imidazo[4,5-c]quinolin-2-yl)methyl)-N-ethylacetamide ≥98%

Supplier: Ambeed
Warning

A1545168-1MG A1545168-10MG A1545168-5MG A1545168-25MG
77598-010EA 32.8 USD
77598-010 77625-368 77625-370 77876-252
N-((4-Amino-7-(1H-pyrazol-5-yl)-3H-imidazo[4,5-c]quinolin-2-yl)methyl)-N-ethylacetamide ≥98%
N-((4-Amino-7-(1H-pyrazol-5-yl)-3H-imidazo[4,5-c]quinolin-2-yl)methyl)-N-ethylacetamide

Precaución: for Research Use Only

Formula: C₁₈H₁₉N₇O
MDL Number: MFCD34567729
CAS Number: 2242952-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR