(3-Methyl-2-(trifluoromethyl)phenyl)boronic acid ≥95%
Supplier: Ambeed
|
Warning
|
A1544901-100MG
A1544901-1G
A1544901-250MG
77876-249EA
183
USD
77876-249
77876-250
77876-251
(3-Methyl-2-(trifluoromethyl)phenyl)boronic acid ≥95%
(3-Methyl-2-(trifluoromethyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₈H₈BF₃O₂ |
MDL Number:
MFCD13194444 CAS Number: 1383576-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...