Sodium 2-(4-(5,7-disulfo-1H-benzo[d]imidazol-2-yl)phenyl)-1H-benzo[d]imidazole-5,7-disulfonate ≥95%

Supplier: Ambeed
Warning

A1541935-100G A1541935-25G A1541935-1G A1541935-5G
77598-002EA 417.18 USD
77598-002 77625-350 77876-238 77876-239
Sodium 2-(4-(5,7-disulfo-1H-benzo[d]imidazol-2-yl)phenyl)-1H-benzo[d]imidazole-5,7-disulfonate ≥95%
Sodium 2-(4-(5,7-disulfo-1H-benzo[d]imidazol-2-yl)phenyl)-1H-benzo[d]imidazole-5,7-disulfonate

Precaución: for Research Use Only

Formula: C₂₀H₁₂N₄Na₂O₁₂S₄
CAS Number: 180898-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR