2-Methyl-1,3-dioxolane-2-ethanamine ≥98%

Supplier: Ambeed
Warning

A154176-1G A154176-250MG
77671-170EA 31 USD
77671-170 77881-205
2-Methyl-1,3-dioxolane-2-ethanamine ≥98%
2-Methyl-1,3-dioxolane-2-ethanamine

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MDL Number: MFCD09923503
CAS Number: 62240-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR