(1R,1'R,2R,2'R)-2,2'-((pentane-1,5-diylbis(oxy))bis(3-oxopropane-3,1-diyl))bis(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium) benzenesulfonate ≥98%

Supplier: Ambeed
Warning

Synonyms: Cisatracurium besylate, (1R,1'R,2R,2'R)-2,2'-((Pentane-1,5-diylbis(oxy))bis(3-oxopropane-3,1-diyl))bis(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium) benzenesulphonate

A153975-1G A153975-100MG
77625-340EA 80.34 USD
77625-340 77879-061
(1R,1'R,2R,2'R)-2,2'-((pentane-1,5-diylbis(oxy))bis(3-oxopropane-3,1-diyl))bis(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium) benzenesulfonate ≥98%
(1R,1'R,2R,2'R)-2,2'-((pentane-1,5-diylbis(oxy))bis(3-oxopropane-3,1-diyl))bis(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium) benzenesulfonate

Precaución: for Research Use Only

Formula: C₆₅H₈₂N₂O₁₈S₂
MDL Number: MFCD00871018
CAS Number: 96946-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR