3-Chloro-4-(4-fluorophenoxy)aniline ≥97%

Supplier: Ambeed
Danger

A153886-1G A153886-250MG
77600-192EA 21.53 USD
77600-192 77671-162
3-Chloro-4-(4-fluorophenoxy)aniline ≥97%
3-Chloro-4-(4-fluorophenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₉ClFNO
MDL Number: MFCD08687313
CAS Number: 937608-56-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR