Bis(4-chlorophenyl)phosphine oxide ≥98%

Supplier: Ambeed
Warning

A1537988-10G A1537988-5G A1537988-1G A1537988-250MG
77625-332EA 475.86 USD
77625-332 77625-334 77876-217 77876-218
Bis(4-chlorophenyl)phosphine oxide ≥98%
Bis(4-chlorophenyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₁₂H₉Cl₂OP
CAS Number: 15948-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR