(1S,3S)-3-((2-Methyl-6-(1-methyl-5-(((methyl(propyl)carbamoyl)oxy)methyl)-1H-1,2,3-triazol-4-yl)pyridin-3-yl)oxy)cyclohexane-1-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A1537897-1MG A1537897-5MG A1537897-10MG A1537897-100MG A1537897-25MG A1537897-50MG
77597-984EA 29.72 USD
77597-984 77597-986 77597-982 77876-214 77876-215 77876-216
(1S,3S)-3-((2-Methyl-6-(1-methyl-5-(((methyl(propyl)carbamoyl)oxy)methyl)-1H-1,2,3-triazol-4-yl)pyridin-3-yl)oxy)cyclohexane-1-carboxylic acid ≥95%
(1S,3S)-3-((2-Methyl-6-(1-methyl-5-(((methyl(propyl)carbamoyl)oxy)methyl)-1H-1,2,3-triazol-4-yl)pyridin-3-yl)oxy)cyclohexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₂H₃₁N₅O₅
MDL Number: MFCD34604078
CAS Number: 2170126-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR