Methyl 2-((methylsulfonyl)methyl)acrylate ≥98%

Supplier: Ambeed
Warning

A1537689-250MG A1537689-5G
77625-328EA 155.53 USD
77625-328 77625-330
Methyl 2-((methylsulfonyl)methyl)acrylate ≥98%
Methyl 2-((methylsulfonyl)methyl)acrylate

Precaución: for Research Use Only

Formula: C₆H₁₀O₄S
CAS Number: 143224-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR