15,16-Dibromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine ≥95%

Supplier: Ambeed
Warning

A1535481-100MG A1535481-1G A1535481-250MG
77876-190EA 40 USD
77876-190 77876-191 77876-192
15,16-Dibromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine ≥95%
15,16-Dibromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine

Precaución: for Research Use Only

Formula: C₁₄H₁₈Br₂O₅
CAS Number: 63862-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR