(((9H-Fluoren-9-yl)methoxy)carbonyl)glycylglycyl-L-phenylalanylglycine ≥95%

Supplier: Ambeed
Warning

Synonyms: Fomc-Gly-Gly-Phe-Gly-OH

A1534320-250MG A1534320-100MG A1534320-1G
77597-976EA 152.73 USD
77597-976 77625-314 77625-316
(((9H-Fluoren-9-yl)methoxy)carbonyl)glycylglycyl-L-phenylalanylglycine ≥95%
(((9H-Fluoren-9-yl)methoxy)carbonyl)glycylglycyl-L-phenylalanylglycine

Precaución: for Research Use Only

Formula: C₃₀H₃₀N₄O₇
MW: 558.58 g/mol
CAS Number: 1817857-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR