5-Bromo-6-fluoroisoindolin-1-one ≥95%

Supplier: Ambeed
Warning

A1534182-100MG A1534182-1G A1534182-250MG
77625-308EA 222.48 USD
77625-308 77625-310 77625-312
5-Bromo-6-fluoroisoindolin-1-one ≥95%
5-Bromo-6-fluoroisoindolin-1-one

Precaución: for Research Use Only

Formula: C₈H₅BrFNO
MDL Number: MFCD23707962
CAS Number: 1427403-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR