3-Amino-N-(benzo[d][1,3]dioxol-5-ylmethyl)-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide ≥98%

Supplier: Ambeed
Warning

A1534053-100MG A1534053-250MG A1534053-25MG A1534053-50MG
77876-168EA 295 USD
77876-168 77876-169 77876-170 77876-171
3-Amino-N-(benzo[d][1,3]dioxol-5-ylmethyl)-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide ≥98%
3-Amino-N-(benzo[d][1,3]dioxol-5-ylmethyl)-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₈H₁₇N₃O₃S
MDL Number: MFCD03668017
CAS Number: 612514-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR