Phenoxathiin-4-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(phenoxathiin-4-yl)-1,3,2-dioxaborolane

A1533562-1G A1533562-250MG
77876-163EA 134 USD
77876-163 77876-164
Phenoxathiin-4-boronic acid pinacol ester ≥98%
Phenoxathiin-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₁₉BO₃S
MW: 326.218 g/mol
MDL Number: MFCD32707006
CAS Number: 2290561-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR