1-(6-(3-Fluoro-5-(trifluoromethyl)benzyl)pyrimidin-4-yl)-N-(2-hydroxyethyl)indoline-4-carboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: PW0787

A1533468-50MG A1533468-100MG
77625-300EA 316.21 USD
77625-300 77625-298
1-(6-(3-Fluoro-5-(trifluoromethyl)benzyl)pyrimidin-4-yl)-N-(2-hydroxyethyl)indoline-4-carboxamide 98+%
1-(6-(3-Fluoro-5-(trifluoromethyl)benzyl)pyrimidin-4-yl)-N-(2-hydroxyethyl)indoline-4-carboxamide

Precaución: for Research Use Only

Formula: C₂₃H₂₀F₄N₄O₂
MW: 460.42 g/mol
MDL Number: MFCD33550902
CAS Number: 2624131-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR