(R)-tert-Butyl 2-(piperazine-1-carbonyl)piperidine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A1533349-1G A1533349-100MG A1533349-250MG
77592-876EA 333.13 USD
77592-876 77625-294 77625-296
(R)-tert-Butyl 2-(piperazine-1-carbonyl)piperidine-1-carboxylate ≥95%
(R)-tert-Butyl 2-(piperazine-1-carbonyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₇N₃O₃
MDL Number: MFCD28000131
CAS Number: 942207-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR