4-[(Dimethylamino)methyl]piperidine ≥95%

Supplier: Ambeed
Danger

A153287-100MG A153287-1G A153287-250MG A153287-5G
77881-184EA 51 USD
77881-184 77881-185 77881-186 77881-187
4-[(Dimethylamino)methyl]piperidine ≥95%
4-[(Dimethylamino)methyl]piperidine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MDL Number: MFCD05215181
CAS Number: 138022-00-1
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR