(11bR)-2,6-Bis(4-fluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%

Supplier: Ambeed
Warning

A1532830-1G A1532830-250MG A1532830-100MG
77597-968EA 208.08 USD
77597-968 77597-970 77625-288
(11bR)-2,6-Bis(4-fluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
(11bR)-2,6-Bis(4-fluorophenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₃₂H₁₉F₂O₄P
MW: 536.46 g/mol
CAS Number: 915038-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR