(11bR)-4-Hydroxy-2,6-diiododinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
Supplier: Ambeed
|
Warning
|
A1532813-250MG
A1532813-100MG
A1532813-1G
77597-966EA
255.23
USD
77597-966
77625-284
77625-286
(11bR)-4-Hydroxy-2,6-diiododinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
(11bR)-4-Hydroxy-2,6-diiododinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
Precaución: for Research Use Only
Formula:
C₂₀H₁₁I₂O₄P MW: 600.08 g/mol |
CAS Number:
861909-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...