2-(5-Bromo-2,3-dihydrobenzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95+%

Supplier: Ambeed
Warning

A1532720-100MG A1532720-1G A1532720-250MG
77876-147EA 47 USD
77876-147 77876-148 77876-149
2-(5-Bromo-2,3-dihydrobenzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95+%
2-(5-Bromo-2,3-dihydrobenzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₁₈BBrO₃
MDL Number: MFCD18783161
CAS Number: 2096333-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR