(E)-6-((6-Chloro-2-methyl-2H-indazol-5-yl)imino)-3-((1-methyl-1H-1,2,4-triazol-3-yl)methyl)-1-(2,4,5-trifluorobenzyl)-1,3,5-triazinane-2,4-dione ≥95%

Supplier: Ambeed
Warning

Synonyms: Ensitrelvir

A1532181-250MG A1532181-5G
77597-958EA 70.73 USD
77597-958 77625-282
(E)-6-((6-Chloro-2-methyl-2H-indazol-5-yl)imino)-3-((1-methyl-1H-1,2,4-triazol-3-yl)methyl)-1-(2,4,5-trifluorobenzyl)-1,3,5-triazinane-2,4-dione ≥95%
(E)-6-((6-Chloro-2-methyl-2H-indazol-5-yl)imino)-3-((1-methyl-1H-1,2,4-triazol-3-yl)methyl)-1-(2,4,5-trifluorobenzyl)-1,3,5-triazinane-2,4-dione

Precaución: for Research Use Only

Formula: C₂₂H₁₇ClF₃N₉O₂
MW: 531.88 g/mol
CAS Number: 2647530-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR