tert-Butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate ≥95%

Supplier: Ambeed
Warning

A1532146-1G A1532146-250MG A1532146-5G
77625-280EA 158.62 USD
77625-280 77876-139 77876-140
tert-Butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate ≥95%
tert-Butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂S₂
MDL Number: MFCD24469377
CAS Number: 535943-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR