AU-15330 ≥98%

Supplier: Ambeed
Warning

Synonyms: (2S,4R)-1-((S)-2-(2-(4-(3-Amino-6-(2-hydroxyphenyl)pyridazin-4-yl)piperazin-1-yl)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide

A1531999-10MG A1531999-5MG A1531999-1MG
77597-954EA 387.45 USD
77597-954 77597-956 77625-276
AU-15330 ≥98%
AU-15330

Precaución: for Research Use Only

Formula: C₃₉H₄₉N₉O₅S
MW: 755.92 g/mol
CAS Number: 2380274-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR