1,3,5-Tris((trimethylsilyl)ethynyl)benzene ≥97%

Supplier: Ambeed
Warning

A1531339-5G A1531339-250MG
77625-272EA 357.41 USD
77625-272 77876-131
1,3,5-Tris((trimethylsilyl)ethynyl)benzene ≥97%
1,3,5-Tris((trimethylsilyl)ethynyl)benzene

Precaución: for Research Use Only

Formula: C₂₁H₃₀Si₃
CAS Number: 18772-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR