3-(2-Cyanopropan-2-yl)-N-(2-fluoro-4-methyl-5-(7-methyl-8-oxo-7,8-dihydropyrido[2,3-d]pyridazin-3-yl)phenyl)benzamide ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
GNE-9815
A1530789-10MG
A1530789-1MG
A1530789-5MG
77597-948EA
369
USD
77597-948
77597-950
77597-952
3-(2-Cyanopropan-2-yl)-N-(2-fluoro-4-methyl-5-(7-methyl-8-oxo-7,8-dihydropyrido[2,3-d]pyridazin-3-yl)phenyl)benzamide ≥98%
3-(2-Cyanopropan-2-yl)-N-(2-fluoro-4-methyl-5-(7-methyl-8-oxo-7,8-dihydropyrido[2,3-d]pyridazin-3-yl)phenyl)benzamide
Precaución: for Research Use Only
Formula:
C₂₆H₂₂FN₅O₂ MW: 455.48 g/mol |
CAS Number:
2729996-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...