N-(2-Aminoethyl)-4-piperidinol ≥98%

Supplier: Ambeed
Danger

Synonyms: 1-(2-aminoethyl)piperidin-4-ol

A153070-1G A153070-250MG
77611-442EA 173.04 USD
77611-442 77685-265
N-(2-Aminoethyl)-4-piperidinol ≥98%
N-(2-Aminoethyl)-4-piperidinol

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O
MW: 144.21 g/mol
MDL Number: MFCD04038390
CAS Number: 129999-60-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR