(2S,4R)-1-((S)-2-(4-Aminobutanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
VH 032 amide-alkylC3-amine
A1530457-50MG
A1530457-250MG
77597-946EA
127.1
USD
77597-946
77625-268
(2S,4R)-1-((S)-2-(4-Aminobutanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride ≥95%
(2S,4R)-1-((S)-2-(4-Aminobutanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride
Precaución: for Research Use Only
Formula:
C₂₆H₃₉Cl₂N₅O₄S MW: 588.59 g/mol |
CAS Number:
2564467-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...