7-Chloro-2-indoleboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A153033-1G A153033-5G
77885-252EA 216 USD
77885-252 77885-253
7-Chloro-2-indoleboronic acid pinacol ester ≥98%
7-Chloro-2-indoleboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BClNO₂
MDL Number: MFCD16295113
CAS Number: 936901-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR