CP-544326 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-((N-(4-(1H-pyrazol-1-yl)benzyl)pyridine-3-sulphonamido)methyl)phenoxy)acetic acid, Taprenepag

A152959-1MG A152959-5MG A152959-10MG
77615-242EA 16.48 USD
77615-242 77615-244 77615-240
CP-544326 ≥98%
CP-544326

Precaución: for Research Use Only

Formula: C₂₄H₂₂N₄O₅S
MW: 478.52 g/mol
MDL Number: MFCD18633300
CAS Number: 752187-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR