Disperse orange 3 ≥95%

Supplier: Ambeed
Warning

Synonyms: 4-((4-Nitrophenyl)diazenyl)aniline

A1529370-1G A1529370-5G
77876-116EA 44 USD
77876-116 77876-117
Disperse orange 3 ≥95%
Disperse orange 3

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₄O₂
MDL Number: MFCD00007856
CAS Number: 730-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR