(4R,4'R)-4,4'-Dimethyl-4,4',5,5'-tetrahydro-2,2'-bioxazole ≥95%

Supplier: Ambeed
Warning

A1529366-1G A1529366-250MG A1529366-100MG A1529366-5G
77625-264EA 364.62 USD
77625-264 77625-266 77876-114 77876-115
(4R,4'R)-4,4'-Dimethyl-4,4',5,5'-tetrahydro-2,2'-bioxazole ≥95%
(4R,4'R)-4,4'-Dimethyl-4,4',5,5'-tetrahydro-2,2'-bioxazole

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
CAS Number: 2755167-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR