2-Allylpent-4-enoic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Valproic Acid Related Compound A

A152913-100MG A152913-1G
77683-285EA 111 USD
77683-285 77683-286
2-Allylpent-4-enoic acid ≥95%, Ambeed.Inc
2-Allylpent-4-enoic acid

Precaución: For research use only.

Formula: C₈H₁₂O₂
MW: 140.17 g/mol
MDL Number: MFCD01709129
CAS Number: 99-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR