Naphthalene-2,6-dicarbaldehyde ≥97%

Supplier: Ambeed
Warning

A152865-1G A152865-250MG
77615-238EA 43.26 USD
77615-238 77748-049
Naphthalene-2,6-dicarbaldehyde ≥97%
Naphthalene-2,6-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₈O₂
MW: 184.19 g/mol
MDL Number: MFCD00462585
CAS Number: 5060-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR