(2-Cyclobutylpyridin-4-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1527991-100MG A1527991-1G A1527991-250MG
77876-103EA 290 USD
77876-103 77876-104 77876-105
(2-Cyclobutylpyridin-4-yl)boronic acid ≥98%
(2-Cyclobutylpyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BNO₂
MDL Number: MFCD22566381
CAS Number: 2225176-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR