(S)-1-(2-Fluorophenyl)ethanamine hydrochloride ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A152657-1G A152657-100MG A152657-250MG A152657-5G
77683-284EA 46 USD
77683-284 77683-283 77892-044 77892-045
(S)-1-(2-Fluorophenyl)ethanamine hydrochloride ≥97%, Ambeed.Inc
(S)-1-(2-Fluorophenyl)ethanamine hydrochloride

Precaución: For research use only.

Formula: C₈H₁₁ClFN
MDL Number: MFCD11865389
CAS Number: 1332832-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR