6-Bromo-3-chloropyrazin-2(1H)-one ≥98%

Supplier: Ambeed
Warning

A1525641-1G A1525641-250MG
77592-862EA 550.43 USD
77592-862 77625-258
6-Bromo-3-chloropyrazin-2(1H)-one ≥98%
6-Bromo-3-chloropyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂O
MW: 209.43 g/mol
MDL Number: MFCD27952472
CAS Number: 1410166-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR