Imidazo[1,5-a]pyridine-1-carbaldehyde ≥97%

Supplier: Ambeed
Warning

A152422-100MG A152422-1G A152422-250MG A152422-5G
77611-428EA 15.45 USD
77611-428 77611-430 77611-432 77611-434
Imidazo[1,5-a]pyridine-1-carbaldehyde ≥97%
Imidazo[1,5-a]pyridine-1-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MDL Number: MFCD06739249
CAS Number: 56671-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR