7-Bromo-4,5-dihydro-1H-pyrido[2,3-e][1,4]diazepin-2(3H)-one ≥95%

Supplier: Ambeed
Warning

A152374-100MG A152374-250MG
77883-193EA 60 USD
77883-193 77883-194
7-Bromo-4,5-dihydro-1H-pyrido[2,3-e][1,4]diazepin-2(3H)-one ≥95%
7-Bromo-4,5-dihydro-1H-pyrido[2,3-e][1,4]diazepin-2(3H)-one

Precaución: for Research Use Only

Formula: C₈H₈BrN₃O
MW: 242.07 g/mol
MDL Number: MFCD11101050
CAS Number: 709650-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR