tert-Butyl 3-(prop-2-yn-1-yloxy)azetidine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A1523595-1G A1523595-5G A1523595-100MG A1523595-250MG
77597-942EA 195.78 USD
77597-942 77625-246 77876-092 77876-093
tert-Butyl 3-(prop-2-yn-1-yloxy)azetidine-1-carboxylate ≥95%
tert-Butyl 3-(prop-2-yn-1-yloxy)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD30726623
CAS Number: 1219606-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR