tert-Butyl 2-(2-hydroxyethyl)-7-azaspiro[3.5]nonane-7-carboxylate ≥95%

Supplier: Ambeed
Warning

A1523594-100MG A1523594-50MG
77625-244EA 281.19 USD
77625-244 77876-091
tert-Butyl 2-(2-hydroxyethyl)-7-azaspiro[3.5]nonane-7-carboxylate ≥95%
tert-Butyl 2-(2-hydroxyethyl)-7-azaspiro[3.5]nonane-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₇NO₃
CAS Number: 1370210-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR