Potassium trifluoro(2-methylprop-1-en-1-yl)borate ≥98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with TBC

A152221-100MG A152221-1G
77892-036EA 67 USD
77892-036 77892-037
Potassium trifluoro(2-methylprop-1-en-1-yl)borate ≥98% stabilized
Potassium trifluoro(2-methylprop-1-en-1-yl)borate

Precaución: for Research Use Only

Formula: C₄H₇BF₃K
MDL Number: MFCD29074005
CAS Number: 1134643-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR