2-(5-((3-Ethoxypyridin-2-yl)oxy)pyridin-3-yl)-N-((3S,5S)-5-fluoropiperidin-3-yl)pyrimidine-5-carboxamide 99% ee 99.9+%

Supplier: Ambeed
Warning

Synonyms: PF-07202954

A1521929-1MG A1521929-10MG A1521929-5MG A1521929-25MG A1521929-50MG
77597-938EA 189.63 USD
77597-938 77625-230 77625-232 77876-074 77876-075
2-(5-((3-Ethoxypyridin-2-yl)oxy)pyridin-3-yl)-N-((3S,5S)-5-fluoropiperidin-3-yl)pyrimidine-5-carboxamide 99% ee 99.9+%
2-(5-((3-Ethoxypyridin-2-yl)oxy)pyridin-3-yl)-N-((3S,5S)-5-fluoropiperidin-3-yl)pyrimidine-5-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₂₃FN₆O₃
MW: 438.45 g/mol
CAS Number: 2639372-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR