(R)-5-Isopropyl-2-mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate ≥97%

Supplier: Ambeed
Danger

A1521103-100MG A1521103-1G A1521103-250MG
77625-224EA 105.06 USD
77625-224 77625-226 77625-228
(R)-5-Isopropyl-2-mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate ≥97%
(R)-5-Isopropyl-2-mesityl-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-2-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₇H₂₄BF₄N₃
MW: 357.2 g/mol
CAS Number: 2216797-99-6
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR