(5aR,10bS)-2-(2,4,6-Triisopropylphenyl)-5a,10b-dihydro-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate ≥95%
Supplier: Ambeed
|
Danger
|
A1521093-50MG
A1521093-100MG
A1521093-1G
A1521093-250MG
77597-930EA
55.35
USD
77597-930
77625-218
77625-220
77625-222
(5aR,10bS)-2-(2,4,6-Triisopropylphenyl)-5a,10b-dihydro-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate ≥95%
(5aR,10bS)-2-(2,4,6-Triisopropylphenyl)-5a,10b-dihydro-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₇H₃₄BF₄N₃O |
CAS Number:
2573798-29-3 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...