(R)-5-Benzyl-2-(4-(tert-butyl)phenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate ≥97%
Supplier: Ambeed
|
Danger
|
A1521082-100MG
A1521082-1G
A1521082-250MG
77625-212EA
77.25
USD
77625-212
77625-214
77625-216
(R)-5-Benzyl-2-(4-(tert-butyl)phenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate ≥97%
(R)-5-Benzyl-2-(4-(tert-butyl)phenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₂H₂₆BF₄N₃O |
UN:
1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...