(R)-5-Benzyl-2-(2,4,6-trichlorophenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate ≥97%

Supplier: Ambeed
Danger

A1521080-250MG A1521080-100MG A1521080-1G
77625-204EA 90.64 USD
77625-204 77876-068 77876-069
(R)-5-Benzyl-2-(2,4,6-trichlorophenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate ≥97%
(R)-5-Benzyl-2-(2,4,6-trichlorophenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₈H₁₅BCl₃F₄N₃O
CAS Number: 2828438-91-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR