rel-Ethyl (1R,8S,9s)-bicyclo[6.1.0]non-4-ene-9-carboxylate ≥95%

Supplier: Ambeed
Warning

A1520967-1G A1520967-250MG
77625-194EA 850.78 USD
77625-194 77625-196
rel-Ethyl (1R,8S,9s)-bicyclo[6.1.0]non-4-ene-9-carboxylate ≥95%
rel-Ethyl (1R,8S,9s)-bicyclo[6.1.0]non-4-ene-9-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
MDL Number: MFCD20264901
CAS Number: 1263291-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR