4-(4,7-Bis(2-(tert-butoxy)-2-oxoethyl)-1,4,7-triazonan-1-yl)-5-(tert-butoxy)-5-oxopentanoic acid ≥98%

Supplier: Ambeed
Warning

A1520867-250MG A1520867-1G
77600-158EA 596.55 USD
77600-158 77876-062
4-(4,7-Bis(2-(tert-butoxy)-2-oxoethyl)-1,4,7-triazonan-1-yl)-5-(tert-butoxy)-5-oxopentanoic acid ≥98%
4-(4,7-Bis(2-(tert-butoxy)-2-oxoethyl)-1,4,7-triazonan-1-yl)-5-(tert-butoxy)-5-oxopentanoic acid

Precaución: for Research Use Only

Formula: C₂₇H₄₉N₃O₈
MDL Number: MFCD29083820
CAS Number: 1190101-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR