N1,N2-Bis(pyridin-2-ylmethyl)oxalamide ≥95%

Supplier: Ambeed
Danger

A1520301-10G A1520301-5G
77625-188EA 745.72 USD
77625-188 77625-190
N1,N2-Bis(pyridin-2-ylmethyl)oxalamide ≥95%
N1,N2-Bis(pyridin-2-ylmethyl)oxalamide

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₄O₂
MW: 270.29 g/mol
MDL Number: MFCD01058969
CAS Number: 2758-86-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR