(4R,5R)-2-(Benzo[b]thiophen-2-yl)-4,5-diphenyl-4,5-dihydrooxazole ≥97%
Supplier: Ambeed
|
Warning
|
A1519933-250MG
A1519933-100MG
A1519933-1G
77625-180EA
148.32
USD
77625-180
77876-046
77876-047
(4R,5R)-2-(Benzo[b]thiophen-2-yl)-4,5-diphenyl-4,5-dihydrooxazole ≥97%
(4R,5R)-2-(Benzo[b]thiophen-2-yl)-4,5-diphenyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₂₃H₁₇NOS |
CAS Number:
2828438-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...